Stampa…

2,5-Dihydroxyphenyl(diphenyl)phosphine oxide ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

D3755-25G D3755-5G
TCIAD3755-25GEA 195 EUR
TCIAD3755-25G TCIAD3755-5G
2,5-Dihydroxyphenyl(diphenyl)phosphine oxide ≥97.0% (by GC)
2,5-Dihydroxyphenyl(diphenyl)phosphine oxide
Formula: C₁₈H₁₅O₃P
Peso molecolare: 310,29 g/mol
CAS Number: 13291-46-8
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae1ee26034f89279ea2c283abec5bfaa6e355189&vid=ae1ee26034f89279ea2c283abec5bfaa6e355189&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 212.0 to 217.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR