Diisononyl cyclohexanedicarboxylate (mixture of isomers)
Fornitore: TCI
Synonyms:
Cyclohexanedicarboxylic Acid Diisononyl Ester
Formula:
C₂₆H₄₈O₄ Peso molecolare: 424,67 g/mol |
CAS Number:
508181-38-2 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | many component mixture |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...