N-[2-(Dimethylamino)ethyl]-N-methylformamide ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
N-(2-(Dimethylamino)ethyl)-N-methylformamide, N-Formyl-N,N',N'-trimethylethylenediamine
D3724-25G
D3724-5G
TCIAD3724-25GEA
149
EUR
TCIAD3724-25G
TCIAD3724-5G
N-[2-(Dimethylamino)ethyl]-N-methylformamide ≥97.0% (by GC, titration analysis)
N-[2-(Dimethylamino)ethyl]-N-methylformamide
Formula:
C₆H₁₄N₂O Peso molecolare: 130,19 g/mol Punto di ebollizione: 96°C Flash Pt: 70°C |
MDL Number:
MFCD04039895 CAS Number: 105669-53-2 |
Risultati del test di specificazione
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...