2'-Deoxycytidine-5'-monophosphate hydrate ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
dCMP, Deoxycytidylic Acid
D3673-1G
D3673-100MG
TCIAD3673-1GEA
283
EUR
TCIAD3673-1G
TCIAD3673-100MG
2'-Deoxycytidine-5'-monophosphate hydrate ≥98.0% (by HPLC, titration analysis)
2'-Deoxycytidine-5'-monophosphate hydrate
Formula:
C₉H₁₄N₃O₇P Peso molecolare: 307,20 g/mol Temperatura di stoccaggio: Refrigerata |
CAS Number:
1032-65-1 EINECS: 213-849-9 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on anh.substance) |
Specific rotation [a]20/D | +35.0 to +43.0 deg (C=1, H2O) (calcd.on anh.substance) |
Water | max. 6.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...