Stampa…

3,5-Dimethyl-1-adamantanamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Memantine Hydrochloride, 1-Amino-3,5-dimethyladamantane Hydrochloride

D3608-25G D3608-5G
TCIAD3608-25GEA 201 EUR
TCIAD3608-25G TCIAD3608-5G
3,5-Dimethyl-1-adamantanamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
3,5-Dimethyl-1-adamantanamine hydrochloride
Formula: C₁₂H₂₁N
Peso molecolare: 215,77 g/mol
Punto di fusione: 292°C
MDL Number: MFCD00214336
CAS Number: 41100-52-1
EINECS: 255-219-6
Merck Index: 14,05806

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR