N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
D3601-1G
D3601-5G
TCIAD3601-1GEA
133
EUR
TCIAD3601-1G
TCIAD3601-5G
N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline ≥98.0% (by GC)
N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Formula:
C₁₆H₂₆BNO₂ |
MDL Number:
MFCD10000956 CAS Number: 920304-57-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...