Stampa…

10,11-Dihydro-5H-dibenzo[b,f]azepine-5-carbonyl chloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2,2'-Iminodibenzyl-5-carbonyl Chloride

D3565-25G D3565-5G
TCIAD3565-25GEA 182 EUR
TCIAD3565-25G TCIAD3565-5G
10,11-Dihydro-5H-dibenzo[b,f]azepine-5-carbonyl chloride ≥98.0% (by HPLC, titration analysis)
10,11-Dihydro-5H-dibenzo[b,f]azepine-5-carbonyl chloride
Formula: C₁₅H₁₂ClNO
Peso molecolare: 257,72 g/mol
MDL Number: MFCD00012314
CAS Number: 33948-19-5
EINECS: 251-756-5
UN: 3261
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Melting point 120.0 to 124.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR