Stampa…

Dimethyl-D-glutamate hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Fornitore: TCI EUROPE

Synonyms: H-D-Glu(OMe)-OMe.HCl, H-D-Glu(OMe)-OMe·HCl, D-Glutamic Acid Dimethyl Ester Hydrochloride

D3560-25G D3560-5G
TCIAD3560-25GEA 120 EUR
TCIAD3560-25G TCIAD3560-5G
Dimethyl-D-glutamate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Dimethyl-D-glutamate hydrochloride
Formula: C₇H₁₃NO₄
Peso molecolare: 211,64 g/mol
Punto di fusione: 91°C
MDL Number: MFCD00235872
CAS Number: 27025-25-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D -24.0 to -28.0 deg (C=2, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR