2,3-Dihydro-1-benzofuran-5-sulphonoylchloride ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
D3553-1G
D3553-5G
TCIAD3553-1GEA
60
EUR
TCIAD3553-1G
TCIAD3553-5G
2,3-Dihydro-1-benzofuran-5-sulphonoylchloride ≥98.0% (by GC)
2,3-Dihydro-1-benzofuran-5-sulphonoylchloride
Formula:
C₈H₇ClO₃S Peso molecolare: 218,66 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD02677703 CAS Number: 115010-11-2 UN: 3261 ADR: 8,II |
Risultati del test di specificazione
Appearance | White to Light yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...