4,10-Dioxatricyclo[5.2.1.0(2,6)]decan-8-en-3-one ≥97.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
4,7-Epoxy-3a,4,7,7a-tetrahydroisobenzofuran-1(3H)-one
D3552-1G
D3552-5G
TCIAD3552-1GEA
53
EUR
TCIAD3552-1G
TCIAD3552-5G
4,10-Dioxatricyclo[5.2.1.0(2,6)]decan-8-en-3-one ≥97.0% (by HPLC)
4,10-Dioxatricyclo[5.2.1.0(2,6)]decan-8-en-3-one
Formula:
C₈H₈O₃ Peso molecolare: 152,15 g/mol |
CAS Number:
72150-22-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Melting point | 91.0 to 95.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...