Stampa…
Diethyl pyrazole-3,5-dicarboxylate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Attenzione
Synonyms: Pyrazole-3,5-dicarboxylic Acid Diethyl Ester, Diethyl 3,5-pyrazoledicarboxylate
D3507-1G D3507-5G
TCIAD3507-1GEA 26 EUR
TCIAD3507-1G TCIAD3507-5G
Diethyl pyrazole-3,5-dicarboxylate ≥98.0% (by GC)
Diethyl pyrazole-3,5-dicarboxylate
Formula: C₉H₁₂N₂O₄
Peso molecolare: 212,21 g/mol
MDL Number: MFCD00152167
CAS Number: 37687-24-4
Visualizza i codici
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 57.0 deg-C