(+)-Di-p-anisoyl-D-tartaric acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(+)-Bis(4-methoxybenzoyl)-D-tartaric Acid, (2S,3S)-2,3-Bis((4-methoxybenzoyl)oxy)succinic acid
D3490-25G
D3490-5G
TCIAD3490-25GEA
83
EUR
TCIAD3490-25G
TCIAD3490-5G
(+)-Di-p-anisoyl-D-tartaric acid ≥98.0% (by HPLC, titration analysis)
(+)-Di-p-anisoyl-D-tartaric acid
Formula:
C₂₀H₁₈O₁₀ Peso molecolare: 418,35 g/mol |
MDL Number:
MFCD07368366 CAS Number: 191605-10-4 EINECS: 471-150-6 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Optical purity (LC) | min. 98.0 ee% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...