6,10-Dibenzyl-N,N'-dimethyl-N,N,N',N'-tetrakis(4-methylbenzyl)-1,4-dioxaspiro[4.5]decane-(2S,3S)-diylbis(methylammonium)tetrafluoroborate [=(S,S)-TaDiAS-2nd] ≥95.0% (by total nitrogen basis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
(S,S)-TaDiAS-2nd
D3476-1G
D3476-200MG
TCIAD3476-1GEA
778
EUR
TCIAD3476-1G
TCIAD3476-200MG
6,10-Dibenzyl-N,N'-dimethyl-N,N,N',N'-tetrakis(4-methylbenzyl)-1,4-dioxaspiro[4.5]decane-(2S,3S)-diylbis(methylammonium)tetrafluoroborate [=(S,S)-TaDiAS-2nd] ≥95.0% (by total nitrogen basis)
6,10-Dibenzyl-N,N'-dimethyl-N,N,N',N'-tetrakis(4-methylbenzyl)-1,4-dioxaspiro[4.5]decane-(2S,3S)-diylbis(methylammonium)tetrafluoroborate [=(S,S)-TaDiAS-2nd]
Formula:
C₅₈H₇₀B₂F₈N₂O₂ Peso molecolare: 1000,82 g/mol |
CAS Number:
2135524-59-1 UN: 1759 ADR: 8,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...