(11S,12S)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₆H₁₆N₂ |
MDL Number:
MFCD06654768 CAS Number: 138517-66-5 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | gt. 156.0 to 160.0 deg-C |
Specific rotation [a]20/D | +17.0 to +22.0 deg (C=1, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...