Dimethyl benzylmalonate ≥95.0% (by GC)
Fornitore: TCI
D3432-25G
D3432-5G
TCIAD3432-25GEA
0
EUR
TCIAD3432-25G
TCIAD3432-5G
Dimethyl benzylmalonate ≥95.0% (by GC)
Dimethyl benzylmalonate
Formula:
C₁₂H₁₄O₄ Punto di ebollizione: 285°C |
MDL Number:
MFCD01445471 CAS Number: 49769-78-0 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...