Stampa…

3,4-Difluorophenylboronic acid (contains varying a mounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,4-Difluorobenzeneboronic acid, (3,4-Difluorophenyl)metaboric acid, (3,4-Difluorophenyl)boronic acid, Dihydroxy(3,4-difluorophenyl)borane

D3350-25G D3350-5G
TCIAD3350-25GEA 92 EUR
TCIAD3350-25G TCIAD3350-5G
3,4-Difluorophenylboronic acid (contains varying a mounts of Anhydride)
3,4-Difluorophenylboronic acid
Formula: C₆H₅BF₂O₂
Peso molecolare: 157,91 g/mol
Punto di fusione: 310°C
MDL Number: MFCD00807405
CAS Number: 168267-41-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR