2,4-Di-tert-pentylphenol ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,4-Di-tert-amylphenol, 2,4-Bis(1,1-dimethylpropyl)phenol, 2,4-Di-tert-pentylphenol
D3294-25G
D3294-5G
TCIAD3294-25GEA
63
EUR
TCIAD3294-25G
TCIAD3294-5G
2,4-Di-tert-pentylphenol ≥98.0% (by GC)
2,4-Di-tert-pentylphenol
Formula:
C₁₆H₂₆O Peso molecolare: 234,38 g/mol Punto di ebollizione: 143°C |
MDL Number:
MFCD00041929 CAS Number: 120-95-6 EINECS: 204-439-0 UN: 3077 ADR: 9,III |
Risultati del test di specificazione
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...