Stampa…

6-(Dibutylamino)-1,3,5-triazine-2,4-dithiol ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: 2-(Dibutylamino)-4,6-dimercapto-1,3,5-triazine

D3265-25G D3265-100G
TCIAD3265-25GEA 21 EUR
TCIAD3265-25G TCIAD3265-100G
6-(Dibutylamino)-1,3,5-triazine-2,4-dithiol ≥98.0% (by HPLC, total nitrogen)
6-(Dibutylamino)-1,3,5-triazine-2,4-dithiol
Formula: C₁₁H₂₀N₄S₂
Peso molecolare: 272,43 g/mol
MDL Number: MFCD00272594
CAS Number: 29529-99-5
EINECS: 249-682-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8249208b28e35ba4668f24fcc696b96c5061057&vid=e8249208b28e35ba4668f24fcc696b96c5061057&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 141.0 to 145.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR