trans-N,N'-Diacetylcyclohexane-1,2-diamine ≥98.0% (by GC)
Fornitore: TCI
D3226-25G
D3226-5G
TCIAD3226-25GEA
190
EUR
TCIAD3226-25G
TCIAD3226-5G
trans-N,N'-Diacetylcyclohexane-1,2-diamine ≥98.0% (by GC)
trans-N,N'-Diacetylcyclohexane-1,2-diamine
Formula:
C₁₀H₁₈N₂O₂ |
MDL Number:
MFCD08276353 CAS Number: 70924-78-6 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...