4-(Dimethylamino)pyridine-N-oxide hydrate ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
DMAPO
D3220-1G
D3220-5G
TCIAD3220-1GEA
135
EUR
TCIAD3220-1G
TCIAD3220-5G
4-(Dimethylamino)pyridine-N-oxide hydrate ≥98.0% (by titrimetric analysis)
4-(Dimethylamino)pyridine-N-oxide hydrate
Formula:
C₇H₁₀N₂O Peso molecolare: 138,17 g/mol Punto di fusione: 97°C |
CAS Number:
1005-31-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystaline |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on anh.substance) |
Water | max. 20.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...