Stampa…

Diethylzinc ∼17% ∼1 mol/L in hexane

Fornitore: TCI EUROPE
Pericolo

D3214-100ML D3214-500ML
TCIAD3214-100MLEA 60 EUR
TCIAD3214-100ML TCIAD3214-500ML
Diethylzinc ∼17% ∼1 mol/L in hexane
Diethylzinc
Formula: C₄H₁₀Zn
Peso molecolare: 123,51 g/mol
MDL Number: MFCD00009021
CAS Number: 557-20-0
EINECS: 209-161-3
UN: 3399
ADR: 4.3,I
Merck Index: 14,03131
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8eecbebedeac8ded014adbe5579efafbce1181f6&vid=a3e8a3de488db8b865732c541b8edfe674f166f0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid to cloudy liquid
Concentration (Chelometric Method) 17.0 to 19.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR