3,6-Di(4-pyridyl)-1,2,4,5-tetrazine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3,6-Di(pyridin-4-yl)-1,2,4,5-tetrazine, 3,6-Bis(4-pyridyl)-1,2,4,5-tetrazine
D3211-1G
D3211-5G
TCIAD3211-1GEA
95
EUR
TCIAD3211-1G
TCIAD3211-5G
3,6-Di(4-pyridyl)-1,2,4,5-tetrazine ≥98.0% (by HPLC, titration analysis)
3,6-Di(4-pyridyl)-1,2,4,5-tetrazine
Formula:
C₁₂H₈N₆ Peso molecolare: 236,24 g/mol |
MDL Number:
MFCD00129054 CAS Number: 57654-36-1 |
Risultati del test di specificazione
Appearance | Yellow to Amber to Dark red powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...