Stampa…

Diammonium ethylenediaminetetraacetate monohydrate ≥98.0% (by total nitrogen and titration analysis)

Fornitore: TCI EUROPE

Synonyms: Diammonium EDTA, Ammonium 2,2'-((2-(bis(carboxymethyl)amino)ethyl)azanediyl)diacetate hydrate, Ethylenediaminetetraacetic Acid Diammonium Salt

D3195-25G D3195-500G
TCIAD3195-25GEA 17 EUR
TCIAD3195-25G TCIAD3195-500G
Diammonium ethylenediaminetetraacetate monohydrate ≥98.0% (by total nitrogen and titration analysis)
Diammonium ethylenediaminetetraacetate monohydrate
Formula: C₁₀H₂₄N₄O₉
Peso molecolare: 344,32 g/mol
MDL Number: MFCD00150463
CAS Number: 304675-80-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Water 4.0 to 7.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR