4,4'-Diamino-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
[2,2'-Bipyridine]-4,4'-diamine, 4,4'-Diamino-2,2'-bipyridine
Formula:
C₁₀H₁₀N₄ |
MDL Number:
MFCD08704188 CAS Number: 18511-69-8 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...