Nickel diethyldithiocarbamate ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Bis(diethyldithiocarbamato)nickel, Diethyldithiocarbamic Acid Nickel Salt
D2879-25G
D2879-500G
TCIAD2879-25GEA
16
EUR
TCIAD2879-25G
TCIAD2879-500G
Nickel diethyldithiocarbamate ≥97.0% (by titrimetric analysis)
Nickel diethyldithiocarbamate
Formula:
C₅H₁₁NS₂ Peso molecolare: 355,22 g/mol |
MDL Number:
MFCD00137706 CAS Number: 14267-17-5 EINECS: 238-157-4 |
Risultati del test di specificazione
Appearance | Green to Dark green powder to crystal |
Purity (Chelometric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...