N,N'-(1,2-Dihydroxyethylene)bisacrylamide ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
N,N'-Diacryloyl-1,2-dihydroxyethylenediamine, 1,2-Bisacrylamido-1,2-ethanediol
D2864-25G
D2864-5G
TCIAD2864-25GEA
231
EUR
TCIAD2864-25G
TCIAD2864-5G
N,N'-(1,2-Dihydroxyethylene)bisacrylamide ≥98.0% (by titrimetric analysis)
N,N'-(1,2-Dihydroxyethylene)bisacrylamide
Formula:
C₈H₁₂N₂O₄ Peso molecolare: 200.19 g/mol Punto di ebollizione: 156°C |
MDL Number:
MFCD00008624 CAS Number: 868-63-3 EINECS: 212-780-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Bromination Method) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...