Stampa…

1,3,2-Dioxathiolane-2,2-dioxide ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1,3,2-Dioxathiolane 2,2-dioxide, Ethylene Sulphate

D2830-5G D2830-25G
TCIAD2830-5GEA 56 EUR
TCIAD2830-5G TCIAD2830-25G
1,3,2-Dioxathiolane-2,2-dioxide ≥98.0% (by GC)
1,3,2-Dioxathiolane-2,2-dioxide
Formula: C₂H₄O₄S
Peso molecolare: 124,11 g/mol
MDL Number: MFCD00221769
CAS Number: 1072-53-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 96.0 to 99.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR