4-Diethylamino-2-butyn-1-ol ≥95.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
D2805-25ML
D2805-5ML
TCIAD2805-25MLEA
422
EUR
TCIAD2805-25ML
TCIAD2805-5ML
4-Diethylamino-2-butyn-1-ol ≥95.0% (by GC, titration analysis)
4-Diethylamino-2-butyn-1-ol
Formula:
C₈H₁₅NO Peso molecolare: 141,21 g/mol Flash Pt: 100°C |
MDL Number:
MFCD00671356 CAS Number: 10575-25-4 EINECS: 234-163-6 |
Risultati del test di specificazione
Appearance | White to Yellow to Orange clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...