2,5-Dichloroamylamine hydrochloride ≥97.0% (by total nitrogen basis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₅H₁₁Cl₂N Punto di fusione: 160°C |
MDL Number:
MFCD00012877 CAS Number: 62922-45-6 EINECS: 263-758-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 159.0 to 163.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...