Stampa…

Disopyramide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(Diisopropylamino)-2-phenyl-2-(2-pyridyl)butyramide

D2793-1G D2793-5G
TCIAD2793-1GEA 46 EUR
TCIAD2793-1G TCIAD2793-5G
Disopyramide ≥98.0% (by HPLC, titration analysis)
Disopyramide
Formula: C₂₁H₂₉N₃O
Peso molecolare: 339,483 g/mol
Punto di fusione: 96°C
MDL Number: MFCD00057366
CAS Number: 3737-09-5
EINECS: 223-110-2
Merck Index: 14,03360

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR