Stampa…

6,6-Diphenylfulvene ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 5-(Diphenylmethylene)-1,3-cyclopentadiene

D2787-1G D2787-5G
TCIAD2787-1GEA 32 EUR
TCIAD2787-1G TCIAD2787-5G
6,6-Diphenylfulvene ≥98.0% (by GC)
6,6-Diphenylfulvene
Formula: C₁₈H₁₄
Peso molecolare: 230,31 g/mol
MDL Number: MFCD00001419
CAS Number: 2175-90-8
EINECS: 218-533-4

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 83.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR