Ethyl-3-(dimethylamino)-3-ethoxy-2,2-difluoropropionate ≥95.0% (by total nitrogen basis)
Fornitore: TCI EUROPE
Synonyms:
3-(Dimethylamino)-3-ethoxy-2,2-difluoropropionic Acid Ethyl Ester
D2782-1G
D2782-5G
TCIAD2782-1GEA
97
EUR
TCIAD2782-1G
TCIAD2782-5G
Ethyl-3-(dimethylamino)-3-ethoxy-2,2-difluoropropionate ≥95.0% (by total nitrogen basis)
Ethyl-3-(dimethylamino)-3-ethoxy-2,2-difluoropropionate
Formula:
C₉H₁₇F₂NO₃ Peso molecolare: 225,24 g/mol Punto di ebollizione: 62°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00722253 CAS Number: 141546-96-5 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (with Total Nitrogen) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...