Stampa…

erythro-5,6-Dodecanediol ≥98.0% (by GC)

Fornitore: TCI

Synonyms: erythro-5,6-Dihydroxydodecane

D2719-100MG D2719-1G
TCIAD2719-100MGEA 264 EUR
TCIAD2719-100MG TCIAD2719-1G
erythro-5,6-Dodecanediol ≥98.0% (by GC)
erythro-5,6-Dodecanediol
Formula: C₁₂H₂₆O₂
Peso molecolare: 202,34 g/mol
MDL Number: MFCD02093467
CAS Number: 70859-32-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e3cee58794aabea33f23c89a1aeda1a51bb1888&vid=7e3cee58794aabea33f23c89a1aeda1a51bb1888&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 119.0 to 124.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR