Stampa…

Dichlorodihexylsilane ≥90.0% (by GC)

Fornitore: TCI
Pericolo

D2712-25G D2712-5G
TCIAD2712-25GEA 233 EUR
TCIAD2712-25G TCIAD2712-5G
Dichlorodihexylsilane ≥90.0% (by GC)
Dichlorodihexylsilane
Formula: C₁₂H₂₆Cl₂Si
Punto di ebollizione: 195°C
Flash Pt: 88°C
MDL Number: MFCD00053203
CAS Number: 18204-93-8
EINECS: 242-093-2
UN: 2987
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR