Stampa…

3,5-Diamino-2,4,6-trimethylbenzenesulphonic acid ≥93.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,6-Diaminomesitylene-4-sulphonic acid, 2,4,6-Trimethyl-3,5-diaminobenzenesulphonic acid

D2709-25G D2709-5G
TCIAD2709-25GEA 103 EUR
TCIAD2709-25G TCIAD2709-5G
3,5-Diamino-2,4,6-trimethylbenzenesulphonic acid ≥93.0% (by titrimetric analysis)
3,5-Diamino-2,4,6-trimethylbenzenesulphonic acid
Formula: C₉H₁₄N₂O₃S
Peso molecolare: 230,28 g/mol
MDL Number: MFCD00043827
CAS Number: 32432-55-6
EINECS: 251-039-7

Visualizza i codici

Risultati del test di specificazione

Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR