Stampa…

(+)-Dipivaloyl-D-tartaric acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI

D2702-1G D2702-5G
TCIAD2702-1GEA 27 EUR
TCIAD2702-1G TCIAD2702-5G
(+)-Dipivaloyl-D-tartaric acid ≥98.0% (by titrimetric analysis)
(+)-Dipivaloyl-D-tartaric acid
Formula: C₁₄H₂₂O₈
MDL Number: MFCD00015634
CAS Number: 76769-55-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 147.0 to 152.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR