2,4-Dimethylaniline-6-sulphonic acid ≥97.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
m-Xylidine-6-sulphonic acid, 2-Amino-3,5-dimethylbenzenesulphonic acid
D2680-25G
D2680-500G
TCIAD2680-25GEA
16
EUR
TCIAD2680-25G
TCIAD2680-500G
2,4-Dimethylaniline-6-sulphonic acid ≥97.0% (by HPLC, titration analysis)
2,4-Dimethylaniline-6-sulphonic acid
Formula:
C₈H₁₁NO₃S Peso molecolare: 201,24 g/mol |
MDL Number:
MFCD00035770 CAS Number: 88-22-2 EINECS: 201-811-4 |
Risultati del test di specificazione
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...