3,5-Di-tert-butylbenzyl alcohol ≥98.0% (by GC)
Fornitore: TCI
Synonyms:
(3,5-Di-tert-butylphenyl)methanol
Formula:
C₁₅H₂₄O |
MDL Number:
MFCD03844785 CAS Number: 77387-57-6 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 59.0 to 63.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...