Stampa…

2,4-Dimethyl-6-hydroxypyrimidine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,6-Dimethylpyrimidin-4-ol, 2,6-Dimethyl-4-pyrimidinol, 6-Hydroxy-2,4-dimethylpyrimidine

D2579-25G D2579-5G
TCIAD2579-25GEA 275 EUR
TCIAD2579-25G TCIAD2579-5G
2,4-Dimethyl-6-hydroxypyrimidine ≥98.0% (by HPLC, titration analysis)
2,4-Dimethyl-6-hydroxypyrimidine
Formula: C₆H₈N₂O
Peso molecolare: 124,14 g/mol
MDL Number: MFCD00006105
CAS Number: 6622-92-0
EINECS: 229-577-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on dried substance)
Melting point 199.0 to 202.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR