Stampa…

Ethyl diphenylphosphonoacetate ≥96.0% (by GC) Horner-Emmons reagent

Fornitore: TCI EUROPE

Synonyms: Diphenylphosphonoacetic Acid Ethyl Ester, Ethyl 2-(diphenoxyphosphoryl)acetate

D2547-1G D2547-5G
TCIAD2547-1GEA 182 EUR
TCIAD2547-1G TCIAD2547-5G
Ethyl diphenylphosphonoacetate ≥96.0% (by GC) Horner-Emmons reagent
Ethyl diphenylphosphonoacetate
Formula: C₁₆H₁₇O₅P
Peso molecolare: 320,28 g/mol
Punto di ebollizione: 220°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD01321165
CAS Number: 16139-79-0

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR