Stampa…

Difluoroacetaldehyde ethyl hemiacetal ≥90.0% (by NMR)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Ethoxy-2,2-difluoroethanol

D2523-25G D2523-5G
TCIAD2523-25GEA 702 EUR
TCIAD2523-25G TCIAD2523-5G
Difluoroacetaldehyde ethyl hemiacetal ≥90.0% (by NMR)
Difluoroacetaldehyde ethyl hemiacetal
Formula: C₄H₈F₂O₂
Peso molecolare: 126,10 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD01321160
CAS Number: 148992-43-2
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ee0e4dc607305f04b6b119cd821cedd24b63a41&vid=2ee0e4dc607305f04b6b119cd821cedd24b63a41&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (NMR) min. 90.0 w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR