Stampa…

4-(Diethylamino)benzoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Diethylaminobenzoic Acid

D2493-25G D2493-5G
TCIAD2493-25GEA 48 EUR
TCIAD2493-25G TCIAD2493-5G
4-(Diethylamino)benzoic acid ≥98.0% (by GC, titration analysis)
4-(Diethylamino)benzoic acid
Formula: C₁₁H₁₅NO₂
Peso molecolare: 193,25 g/mol
MDL Number: MFCD00002538
CAS Number: 5429-28-7
EINECS: 226-582-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81bc2bfcfca821d332c2cc405259def60496de8f&vid=81bc2bfcfca821d332c2cc405259def60496de8f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Green to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 193.0 to 197.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR