Stampa…

3,5-Diisopropylpyrazole ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 3,5-Diisopropyl-1H-pyrazole

D2407-25G D2407-5G
TCIAD2407-25GEA 457 EUR
TCIAD2407-25G TCIAD2407-5G
3,5-Diisopropylpyrazole ≥98.0% (by GC, titration analysis)
3,5-Diisopropylpyrazole
Formula: C₉H₁₆N₂
MDL Number: MFCD00671516
CAS Number: 17536-00-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 87.0 to 92.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR