2,7-Di-tert-butylnaphthalene ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₈H₂₄ |
MDL Number:
MFCD00094397 CAS Number: 10275-58-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 102.0 to 106.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...