Stampa…

Dimethyldipalmitylammonium bromide ≥97.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: Dihexadecyldimethylammonium Bromide, Dicetyldimonium bromide, N-Hexadecyl-N,N-dimethylhexadecan-1-aminium bromide

D2354-25G D2354-500G
TCIAD2354-25GEA 54 EUR
TCIAD2354-25G TCIAD2354-500G
Dimethyldipalmitylammonium bromide ≥97.0% (by titrimetric analysis)
Dimethyldipalmitylammonium bromide
Formula: C₃₄H₇₂BrN
Peso molecolare: 574.86 g/mol
MDL Number: MFCD00191428
CAS Number: 70755-47-4
EINECS: 274-849-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Solubility in hot Methanol almost transparency
Water max. 3.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR