Stampa…

4-Methyl-N-phenyl-N-(p-tolyl)aniline ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N,N-Di-p-tolylaniline, Phenyl(di-p-tolyl)amine, 4,4-Dimethyltriphenylamine, Bis(4-methylphenyl)phenylamine, 4,4'-Dimethyltriphenylamine

D2347-1G D2347-25G D2347-5G
TCIAD2347-1GEA 43 EUR
TCIAD2347-1G TCIAD2347-25G TCIAD2347-5G
4-Methyl-N-phenyl-N-(p-tolyl)aniline ≥98.0% (by GC)
4-Methyl-N-phenyl-N-(p-tolyl)aniline
Formula: C₂₀H₁₉N
Peso molecolare: 273,3700 g/mol
MDL Number: MFCD00145004
CAS Number: 20440-95-3
EINECS: 243-822-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 108.0 to 111.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR