Stampa…

tert-Butyl-2-((diphenylmethylene)amino)acetate ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: N-(Diphenylmethylene)glycine tert-butyl ester, Glycine, N-(diphenylmethylene)-, 1,1-dimethylethyl ester, tert-Butyl 2-((diphenylmethylene)amino)acetate

D2322-1G D2322-25G D2322-5G
TCIAD2322-1GEA 18 EUR
TCIAD2322-1G TCIAD2322-25G TCIAD2322-5G
tert-Butyl-2-((diphenylmethylene)amino)acetate ≥98.0% (by HPLC, total nitrogen)
tert-Butyl-2-((diphenylmethylene)amino)acetate
Formula: C₁₉H₂₁NO₂
Peso molecolare: 295,382 g/mol
MDL Number: MFCD00134280
CAS Number: 81477-94-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 112.0 to 116.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR