Stampa…

2,5-Dibromohexane mixture of diasteroisomers ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

D2298-25G D2298-5G
TCIAD2298-25GEA 156 EUR
TCIAD2298-25G TCIAD2298-5G
2,5-Dibromohexane mixture of diasteroisomers ≥98.0% (by GC)
2,5-Dibromohexane
Formula: H₃CBrCH(CH₂)₂BrCHCH₃
Peso molecolare: 243,97 g/mol
MDL Number: MFCD00039177
CAS Number: 24774-58-1
EINECS: 246-457-1

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 % (total of diastereoisomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR