trans-2,3-Dichloro-1,4-dioxane ≥96.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
trans-2,3-Dichloro-1,4-diethylene Dioxide
D2279-25G
D2279-5G
TCIAD2279-25GEA
170
EUR
TCIAD2279-25G
TCIAD2279-5G
trans-2,3-Dichloro-1,4-dioxane ≥96.0% (by GC)
trans-2,3-Dichloro-1,4-dioxane
Formula:
C₄H₆Cl₂O₂ Peso molecolare: 156,99 g/mol Punto di ebollizione: 82°C |
MDL Number:
MFCD00191421 CAS Number: 3883-43-0 UN: 2811 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...