Chlorodiethylisopropylsilane ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Diethylisopropylsilyl Chloride
D2262-1G
D2262-5G
TCIAD2262-1GEA
44
EUR
TCIAD2262-1G
TCIAD2262-5G
Chlorodiethylisopropylsilane ≥97.0% (by titrimetric analysis)
Chlorodiethylisopropylsilane
Formula:
C₇H₁₇ClSi Peso molecolare: 164,75 g/mol Punto di ebollizione: 75°C Flash Pt: 38°C |
MDL Number:
MFCD00191631 CAS Number: 107149-56-4 UN: 2986 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...